Pipette stand, rotary
Supplier: Brand
30900
BRND30900EA 181 SGD
BRND30900
Pipette stand, rotary
Pipette Stands
Rotary pipette stand made of PP.
  • For up to 94 graduated or calibrated pipettes of different sizes
  • Rotary model with additional lower support plate to protect pipette tips
  • Slightly raised edge of base retains liquid residues
Order Now
SPECIFICATIONS
Environmentally Preferable