Volumetric flasks for oil content determination, white markings, SILBERBRAND
Supplier: Brand
365538
BRND365538EA 394 SGD
BRND365538
Volumetric flasks for oil content determination, white markings, SILBERBRAND
Flasks Volumetric Flasks
Borosilicate glass 3.3, with NS hollow glass stopper.
  • Calibrated to contain (TC, In)
  • Subdivision 0 to 5 ml (Error limit 0,10 ml ±)
  • Subdivision 5 to 30 ml (Error limit 0,5 ml ±)
For determination of oil content in aqueous oil emulsions (e.g. drilling oil).
Certifications: DIN 51368.