Dissolved oxygen bottles
Supplier: Brand
386038
BRND386038EA 87.4 SGD
BRND386038
Dissolved oxygen bottles
Bottles Dissolved Oxygen Bottles
Soda-lime glass.
  • Solid, obliquely cut standard-ground glass stopper can be secured with a fastening clip
  • Each bottle is calibrated with its own stopper; hence, stoppers and bottles are not interchangeable
  • Each bottle and its stopper is marked with a unique matching identification number
For the determination of oxygen dissolved water. The measured volume is indicated to ± 0,01 ml. White labelling area.
Order Now
SPECIFICATIONS
Environmentally Preferable