Desiccators, with lid
Supplier: Brand
65805 65810 65815
BRND65805EA 277 SGD
BRND65805 BRND65810 BRND65815
Desiccators, with lid
Desiccators Desiccators, Plastic
Dessicators with polycarbonate lid, PP base and desiccant tray. Venting stopper with non-return valve in lid. Seal between lid and base: polychlorobutadiene rubber (CK) O-ring, compressed by vacuum.
  • Light-weight and easy to handle
Order Now
SPECIFICATIONS
Environmentally Preferable