Desiccator plates
Supplier: Brand
66015 66025 66030 65965 65975 65980
BRND66015EA 36.1 SGD
BRND66015 BRND66025 BRND66030 BRND65965 BRND65975 BRND65980
Desiccator plates
Desiccators Desiccators, Plates
Desiccator plates for glass and plastic desiccators.
  • Central hole measures approximately 20 mm diameter
  • Small perforations measure approximately 5 mm diameter