Microplates, 96-well
Supplier: Brand
701330
BRND701330EA 395 SGD
BRND701330
Microplates, 96-well
Microplates HTS Microplates
PP, round bottom wells, non sterile.
  • Compatible with virtually all leading microplate centrifuges
  • Raised well rims minimise cross-contamination
  • Plates can be sealed using self-adhesive films, such as DMSO-resistant sealing film (cross-cut) with alphanumeric coding
For volumes up to 300 µl.