Counting chambers, BLAUBRAND® Neubauer Improved
Supplier: Brand
717810
BRND717810EA 348 SGD
BRND717810
Counting chambers, BLAUBRAND® Neubauer Improved
Slides Counting Chambers
This counting chamber feature the same ruling as improved Neubauer but with a rhodium coated chamber bottom.
  • Rulings engraved into the rhodium layer appear bright under normal microscope settings
  • By altering the contrast, the microscope image can be reversed
  • Rulings can appear brighter or darker, as required