Counting chambers, BLAUBRAND® Thoma
Supplier: Brand
718005 718020
BRND718005EA 170 SGD
BRND718005 BRND718020
Counting chambers, BLAUBRAND® Thoma
Slides Counting Chambers
Double ruling. The net division corresponds to the large square lying in the middle of the Neubauer chamber. The area of the small square is 0,0025 mm². Since the outer large squares are not included, the Thoma system is only used to count thrombocytes and erythrocytes.