Urine beaker, with press-on lids
Supplier: Brand
758901 758902
BRND758901EA 326 SGD
BRND758901 BRND758902
Urine beaker, with press-on lids
Clinical Diagnostic Supplies Clinical Sample Collection/Preparation Urine Sample Collection/Preparation
PP, with red PE lid.
  • Lid is simply sealed by being pressed on
  • Easy to use
  • Simply cut off integrated spout tip and insert test strip
For hygienic and odorless urine sample handling.