Disposable cuvettes
Supplier: Brand
759150 759220 759243 759242 759241 759240 759230 759210 759200 759170 759215 759235
BRND759150EA 142 SGD
BRND759150 BRND759220 BRND759243 BRND759242 BRND759241 BRND759240 BRND759230 BRND759210 BRND759200 BRND759170 BRND759215 BRND759235
Disposable cuvettes
Cuvettes Spectrophotometer Cuvettes
Disposable cuvettes often replace expensive and sensitive cuvettes made from glass and quartz.
  • High chemical resistance for use with polar solvents, acids and caustic solutions
  • Measurements at 260 or 280 nm and in the visible wavelength range
  • Round coloured caps, for microcuvettes only, ensure easy sample identification and provide a tight seal to –20 °C
Suitable for photometric measurements of proteins, DNA and RNA in the UV range.
Packaging: Individually wrapped micro UV cuvettes are free of DNase, DNA and RNase.
Order Now
SPECIFICATIONS
Environmentally Preferable