Rack for cuvettes
Supplier: Brand
759500
BRND759500EA 37.2 SGD
BRND759500
Rack for cuvettes
Racks Cuvette Racks
PP.
  • Numbered positions to aid sample identification
  • Autoclavable at 121 °C
Rack with numbered positions for 2×8 cuvettes of 10 mm path length.