Racks for disposal bags
Supplier: Brand
759703
BRND759703EA 36.2 SGD
BRND759703
Racks for disposal bags
Racks Bag Racks
These bags are made of epoxy-coated steel wire, with rubber feet.
  • Not autoclavable
  • Inner diameter: 120 mm
  • Height: 250 mm