Centrifuge tubes
Supplier: Brand
778012
BRND778012EA 211 SGD
BRND778012
Centrifuge tubes
Tubes Centrifuge Tubes
AR® glass, conical bottom, beaded rim (except 525-0500 cone-pointed bottom, rimless).
  • Withstands forces up to 3000×g