Microcentrifuge tubes, 0,5 ml
Supplier: Brand
780507
BRND780507EA 156 SGD
BRND780507
Microcentrifuge tubes, 0,5 ml
Tubes Microcentrifuge Tubes
PP, autoclavable with attached cap.
  • Can withstand centrifugation up to 10000×g
  • Flat cap can be easily labelled and pierced
  • Tight fitting attached cap provides leak free seal, yet re-opens easily