Microcentrifuge tubes
Supplier: Brand
780540 780536 780546
BRND780540EA 143 SGD
BRND780540 BRND780536 BRND780546
Microcentrifuge tubes
Tubes Microcentrifuge Tubes
PP, translucent.
  • With lid locking
  • Frosted marking area
  • Subdivisions for approx. volume determination
  • Free of endotoxins, DNase and Rnase