Microcentrifuge tubes, 2,0 ml
Supplier: Brand
780550
BRND780550EA 103 SGD
BRND780550
Microcentrifuge tubes, 2,0 ml
Tubes Microcentrifuge Tubes
PP, graduated, autoclavable with attached cap.
  • Can withstand centrifugation up to 20000×g
  • Flat cap can be easily labelled and pierced
  • Frosted marking area