Microcentrifuge tubes with attached screw caps, 0,5/1,5/2,0 ml
Supplier: Brand
780710 780711 780712 780713 780714
BRND780710EA 798 SGD
BRND780710 BRND780711 BRND780712 BRND780713 BRND780714
Microcentrifuge tubes with attached screw caps, 0,5/1,5/2,0 ml
Tubes Microcentrifuge Tubes
PP, graduated, autoclavable, PP screw cap with silicone seal.
  • Can withstand centrifugation to 17000×g
  • Ideal for storage of serum and blood samples as well as for boiling and centrifugation
  • Positive sealing screw cap with silicone seal
  • Fast opening and closing with 1¹/₄ turn of cap
  • Operating range: −196 to +121 °C, suitable for freezing samples in the gas phase of liquid nitrogen at −196 °C
*without graduations
Ordering information: Coloured cap inserts for identification are available separately.