Microcentrifuge tubes, 0,5/1,5/2,0 ml
Supplier: Brand
780730 780731 780732 780733 780734 780740 780741 780742 780743 780744 780750 780751 780752 780753 780754
BRND780730EA 407 SGD
BRND780730 BRND780731 BRND780732 BRND780733 BRND780734 BRND780740 BRND780741 BRND780742 BRND780743 BRND780744 BRND780750 BRND780751 BRND780752 BRND780753 BRND780754
Microcentrifuge tubes, 0,5/1,5/2,0 ml
Tubes Microcentrifuge Tubes
PP, graduated, sterile with PP screw cap or non sterile without PP screw cap, autoclavable.
  • Can withstand centrifugation to 17000×g
  • Cap with silicone seal
  • With frosted marking area
  • Sterile tubes are DNA, DNase, and RNase-free, endotoxin-free, non-mutagenic, non-toxic
Temperature range: −196 to +121 °C.
*without graduations.