Microcentrifuge tubes, 0,5/1,5/2,0 ml
Supplier: Brand
780757 780756 780758 780759 780760 780761 780762 780773 780774 780775 780755 780763 780764 780770 780771 780772
BRND780757EA 909 SGD
BRND780757 BRND780756 BRND780758 BRND780759 BRND780760 BRND780761 BRND780762 BRND780773 BRND780774 BRND780775 BRND780755 BRND780763 BRND780764 BRND780770 BRND780771 BRND780772
Microcentrifuge tubes, 0,5/1,5/2,0 ml
Tubes Microcentrifuge Tubes
PP, graduated, sterile or non sterile, with or without tamper-evident screw cap, autoclavable.
  • Can withstand centrifugation to 17000×g
  • DNA, DNase and RNase free, non-mutagenic, non-toxic
  • With frosted marking area
Operating range: −196 to +121 °C
Packaging: Pack of 500 sterile or 1000 non sterile version and caps.