Sealing film, self-adhesive, for cold storage
Supplier: Brand
781381
BRND781381EA 437 SGD
BRND781381
Sealing film, self-adhesive, for cold storage
Seals Microplate Seals
Aluminium sealing film can be pierced easily with a pipette tip for sample removal. Temperature range: −80 to +120 °C.