Sealing film, self-adhesive for PCR, real time PCR
Supplier: Brand
781383
BRND781383EA 254 SGD
BRND781383
Sealing film, self-adhesive for PCR, real time PCR
Seals Microplate Seals
Polyester flims suitable for PCR and Real-Time PCR measurements. Temperature range: –40 °C to +120 °C.