Sealing film, self-adhesive for ELISA, PCR
Supplier: Brand
781390
BRND781390EA 415 SGD
BRND781390
Sealing film, self-adhesive for ELISA, PCR
Seals Microplate Seals
Transparent PP film allows the visual inspection of the samples. Temperature range: −40 to +125 °C.