Sealing film, self-adhesive for ELISA, Real-Time PCR (qPCR)
Supplier: Brand
781391
BRND781391EA 600 SGD
BRND781391
Sealing film, self-adhesive for ELISA, Real-Time PCR (qPCR)
Seals Microplate Seals
Highly transparent film made of polyester is suitable for Real-Time PCR measurements. Temperature range: −40 to +120 °C.