PCR plates for qPCR, 24-well and 48-well
Supplier: Brand
781411 781415 781412 781416
BRND781411EA 225 SGD
BRND781411 BRND781415 BRND781412 BRND781416
PCR plates for qPCR, 24-well and 48-well
Microplates PCR Plates
PP, non skirted. Manufactured under cleanroom conditions, and made from pure, high quality materials (without additives) these PCR plates are suitable for use with most commercially available thermal cyclers.
  • Ultra-thin walled design to facilitate constant, rapid and precise heat transfer
  • Smooth vessel interior minimises the binding of enzymes and nucleic acid to the walls
  • Alphanumeric coding embossed and stamped in blue
  • Raised well rims protect against cross-contamination and provide a tight seal in combination with matching sealing mat
  • Free from DNase,
More Product Information