1536-well (UHTS) microplates, non treated, BRANDplates®
Supplier: Brand
781640 781641 781642
BRND781640EA 2640 SGD
BRND781640 BRND781641 BRND781642
1536-well (UHTS) microplates, non treated, BRANDplates®
Microplates HTS Microplates
These PS flat bottom plates are available sterile or non sterile.
  • All BRANDplates® are alphanumerically labelled, stackable and compatible with most standard microplate readers and washers
  • White plates provide maximum reflection and minimum crosstalk during luminescence measurements; black plates provide minimum back light scatter during fluorescence analyses
  • Sterile plates are supplied individually wrapped with lid
pureGrade™: Non treated, medium binding, non sterile surface. These plates are suitable for homogeneous assays, screening and storage.