96-Well microplates, non treated, BRANDplates®
Supplier: Brand
781600 781601 781602 781603 781604 781605 781607 781608 781610 781611 781660 781661 781662 781663 781665 781668 781670 781671 781614
BRND781600EA 328 SGD
BRND781600 BRND781601 BRND781602 BRND781603 BRND781604 BRND781605 BRND781607 BRND781608 BRND781610 BRND781611 BRND781660 BRND781661 BRND781662 BRND781663 BRND781665 BRND781668 BRND781670 BRND781671 BRND781614
96-Well microplates, non treated, BRANDplates®
Microplates HTS Microplates
PS, solid plates or with transparent base. Plates are available with different well bottom shapes (round U-bottom, conical V-bottom, flat F-bottom or C flat bottom with curved edges), in different colours, and either sterile or non sterile.
  • All BRANDplates® are alphanumerically labelled, stackable and compatible with most standard microplate readers and washers
  • White plates provide maximum reflection and minimum crosstalk during luminescence measurements; black plates provide minimum back light scatter during fluorescence analysis
  • Sterile plates are
More Product Information