384-well microplates for immunology, BRANDplates®
Supplier: Brand
781740 781741 781742
BRND781740EA 875 SGD
BRND781740 BRND781741 BRND781742
384-well microplates for immunology, BRANDplates®
Microplates Immunology Microplates and Strips
PS, solid or with transparent base. These flat bottom plates are available in different colours, and with standard or transparent base formats.
immunoGrade™: The surface of these plates is optimised for the immobilisation of IgG, showing a high binding capacity for molecules with hydrophilic and hydrophobic regions. Plates with an immunoGrade™ surface are the best choice for standard ELISA applications.
The surface of these plates is optimised for the immobilisation of IgG, showing a high binding capacity for molecules with hydrophilic and hydrophobic regions. Plates with anMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable