96-well microplates for immunology, BRANDplates®
Supplier: Brand
781720 781721 781722 781724 781731 781780 781782 781840 781842
BRND781720EA 534 SGD
BRND781720 BRND781721 BRND781722 BRND781724 BRND781731 BRND781780 BRND781782 BRND781840 BRND781842
96-well microplates for immunology, BRANDplates®
Microplates Immunology Microplates and Strips
PS, solid or with transparent base. Plates are available with different well shapes (round U-bottom, conical V-bottom, flat F-bottom or C-flat bottom with curved edges), in different colours, and with a choice of treated surfaces.
  • All BRANDplates® are alphanumerically labelled, stackable and compatible with most standard microplate readers and washers
  • Standard 96-well plates have coloured alphanumeric code identifying surface type (cell culture treated orange, immuno treated blue, non-treated grey)
  • White plates provide maximum reflection and minimum
More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable