Cell culture inserts, 2 in 1, cellGrade™ plus
Supplier: Brand
782700
BRND782700EA 1070 SGD
BRND782700
Cell culture inserts, 2 in 1, cellGrade™ plus
Microplates Insert Plates and Inserts
Cell culture inserts with cellGrade™ plus treated membranes for improved cell attachment and growth.
  • Can be used as a hanging insert (fits nearly all multiwell plates) or, for bigger wells and cuture dishes, as a standing insert
  • cellGrade™ plus surface for excellent cell attachment and growth
  • Highly transparent membranes give good visibility of cell cultures
  • High pore density contributes to good permeability
  • Innovative packaging provides for efficient unpacking and reduces waste
Ordering information: Inserts are available for 6-, 12- and 24-well plateMore Product Information