Sample dippers
Supplier: Brand
90438 90448 90454 90462
BRND90438EA 531 SGD
BRND90438 BRND90448 BRND90454 BRND90462
Sample dippers
Samplers Sample Dippers
Sampling dipper with reinforced rim and spout.
  • PTFE handle with steel core
Optional extension rod for increased handle length.
Order Now
SPECIFICATIONS
Environmentally Preferable