Electrodes kit, 2-Needle Array™
Supplier: BTX
45-0167
BTXI45-0167EA 2390 SGD
BTXI45-0167
Electrodes kit, 2-Needle Array™
Electrodes Electroporation Electrodes
These in vivo style electrode specifically designed for muscle gene delivery. Among the non-viral techniques for in vivo gene transfer, the direct injection of plasmid DNA into muscle is simple, inexpensive and safe. The electrode is available in two gap sizes, 5 mm and 10 mm.
  • Handle made of Delrin, needle made of stainless steel
  • 5 mm 2-Needle array and handle is designed for small muscle masses such as mouse tibialis
  • 10 mm 2-Needle array and handle is designed for larger muscle masses such as rat gastrocnemius
Generator compatibility: ECM 830 and ECMMore Product Information