Soxhlet extractors, flanged extractor head
Supplier: BUCHI
011744 011745
BUCH011744EA 9450 SGD
BUCH011744 BUCH011745
Soxhlet extractors, flanged extractor head
Extractors Soxhlet and Soxhlet-Type Extractors
Borosilicate glass.
  • Excellent chemical resistance
  • Excellent temperature resistance