Glass drying oven, B-585TO
Supplier: BUCHI
037010 037133 046600
BUCH037010EA 2830 SGD
BUCH037010 BUCH037133 BUCH046600
Glass drying oven, B-585TO
Evaporators Microevaporators
Small drying oven with metal housing and robust platform; glass drying chamber for optimal monitoring of the sample. Solvent residues can be dried up to a maximum temperature of 300 °C.
  • Quick drying of small quantities
  • Small footprint
  • Option to perform sublimation
  • Graphic display with manual and programmable mode
  • Overtemperature protection
  • Glass bulb at the end to contain desiccating agents outside the oven
Vacuum connection option, optional automatic on/off control with a Büchi pump connected.
Neutral gas flushing option to avoid oxidising theMore Product Information