Micro-evaporator, glass oven, B-585 Kugelrohr
Supplier: BUCHI
046601
BUCH046601EA 33010 SGD
BUCH046601
Micro-evaporator, glass oven, B-585 Kugelrohr
Evaporators Microevaporators
For drying, distillation, dry freezing and sublimation of small quantities. Ideal for fractional distillation of small quantities of solvents up to 300 °C.
  • Visual monitoring of operation
  • Integral condenser
  • Digital display with continuous temperature control
  • Vacuum and/or inert gas connection
  • Overtemperature protection and contact safeguard on glass steam circulation tube
Ordering information: Device supplied complete with 1 rotation drying flask (60 ml) with vapor duct (DDR), 3 ball tubes (20 ml) with vapor duct, 1 cooling device, diverse standardMore Product Information