Rotary evaporator, large scale, Rotavapor® R-250 EX
Supplier: BUCHI
8621D551
BUCH8621D551EA 0 SGD
BUCH8621D551
Rotary evaporator, large scale, Rotavapor® R-250 EX
Evaporators Rotary Evaporators
The Rotavapor R-250 EX is the EX rated version of the Rotavapor R-250. It fulfills the high requirements of the ATEX regulations and fits perfect to your Ex-proof production laboratory. It is designed for easy and safe operation combined with high performance.
  • Intuitive operation and handling guarantee safe and efficient industrial distillation
  • Immediate shut-down due to safety cut-off
  • Safe handling and spring-loaded valves protect the glass
  • Easy flask handling due to the unique flask coupling
  • Long life time due to exclusive use of corrosion-free stainless
More Product Information