Parallel vortex evaporators, Multivapor™ P-6
Supplier: BUCHI
MP21000C00 MP21000C10 MP21000C20 MP21000R00 MP21000S00 MP21000S10 MP21000S20 MP21001C00 MP21001C10 MP21001C20 MP21001S00 MP21001S20 MP21099C00 MP21099C10 MP21099C20 MP21099R00 MP21099S00 MP21099S20 MP21100C00 MP21100C10 MP21100R00 MP21100S00 MP21100S10 MP21100S20 MP21101C00 MP21101C10 MP21101C20 MP21101R00 MP21101S00 MP21101S10 MP21199C00 MP21199C10 MP21199C20 MP21199R00 MP21199S00 MP21199S10 MP21199S20
BUCHMP21000C00EA 0 SGD
BUCHMP21000C00 BUCHMP21000C10 BUCHMP21000C20 BUCHMP21000R00 BUCHMP21000S00 BUCHMP21000S10 BUCHMP21000S20 BUCHMP21001C00 BUCHMP21001C10 BUCHMP21001C20 BUCHMP21001S00 BUCHMP21001S20 BUCHMP21099C00 BUCHMP21099C10 BUCHMP21099C20 BUCHMP21099R00 BUCHMP21099S00 BUCHMP21099S20 BUCHMP21100C00 BUCHMP21100C10 BUCHMP21100R00 BUCHMP21100S00 BUCHMP21100S10 BUCHMP21100S20 BUCHMP21101C00 BUCHMP21101C10 BUCHMP21101C20 BUCHMP21101R00 BUCHMP21101S00 BUCHMP21101S10 BUCHMP21199C00 BUCHMP21199C10 BUCHMP21199C20 BUCHMP21199R00 BUCHMP21199S00 BUCHMP21199S10 BUCHMP21199S20
Parallel vortex evaporators, Multivapor™ P-6
Evaporators Parallel Synthesis Systems
This compact instrument for parallel evaporation of up to six samples under vacuum conditions is designed to optimise existing personal process workflows such as work-up of synthesys samples, evaporation of extraction samples or chromatographic fractions.
  • Small footprint: Requires minimal bench top space, even when combined with the Rotavapor®
  • Samples are visible throughout the whole evaporation process
  • Temperature and orbital movement settings are adjustable, setpoint and actual temperature are shown on a display
  • Environmental friendly and safe:
More Product Information
Order Now
SPECIFICATIONS
Model Multivapor P-6 Multivapor P-6 Multivapor P-6 Multivapor P-6 Multivapor P-6 Multivapor P-6 Multivapor P-6 Multivapor P-6 Multivapor P-6 Multivapor P-6 Multivapor P-6 Multivapor P-6 Multivapor P-6 Multivapor P-6 Multivapor P-6 Multivapor P-6 Multivapor P-6 Multivapor P-6 Multivapor P-6 Multivapor P-6 Multivapor P-6 Multivapor P-6 Multivapor P-6 Multivapor P-6 Multivapor P-6 Multivapor P-6 Multivapor P-6 Multivapor P-6 Multivapor P-6 Multivapor P-6 Multivapor P-6 Multivapor P-6 Multivapor P-6 Multivapor P-6 Multivapor P-6 Multivapor P-6 Multivapor P-6
Vibration frequency 0 - 370 min⁻¹ 0 - 370 min⁻¹ 0 - 370 min⁻¹ 0 - 370 min⁻¹ 0 - 370 min⁻¹ 0 - 370 min⁻¹ 0 - 370 min⁻¹ 0 - 370 min⁻¹ 0 - 370 min⁻¹ 0 - 370 min⁻¹ 0 - 370 min⁻¹ 0 - 370 min⁻¹ 0 - 370 min⁻¹ 0 - 370 min⁻¹ 0 - 370 min⁻¹ 0 - 370 min⁻¹ 0 - 370 min⁻¹ 0 - 370 min⁻¹ 0 - 370 min⁻¹ 0 - 370 min⁻¹ 0 - 370 min⁻¹ 0 - 370 min⁻¹ 0 - 370 min⁻¹ 0 - 370 min⁻¹ 0 - 370 min⁻¹ 0 - 370 min⁻¹ 0 - 370 min⁻¹ 0 - 370 min⁻¹ 0 - 370 min⁻¹ 0 - 370 min⁻¹ 0 - 370 min⁻¹ 0 - 370 min⁻¹ 0 - 370 min⁻¹ 0 - 370 min⁻¹ 0 - 370 min⁻¹ 0 - 370 min⁻¹ 0 - 370 min⁻¹
Environmentally Preferable
Heating power max. 800 W max. 800 W max. 800 W max. 800 W max. 800 W max. 800 W max. 800 W max. 800 W max. 800 W max. 800 W max. 800 W max. 800 W max. 800 W max. 800 W max. 800 W max. 800 W max. 800 W max. 800 W max. 800 W max. 800 W max. 800 W max. 800 W max. 800 W max. 800 W max. 800 W max. 800 W max. 800 W max. 800 W max. 800 W max. 800 W max. 800 W max. 800 W max. 800 W max. 800 W max. 800 W max. 800 W max. 800 W
Temperature range Ambient…+95 °C Ambient…+95 °C Ambient…+95 °C Ambient…+95 °C Ambient…+95 °C Ambient…+95 °C Ambient…+95 °C Ambient…+95 °C Ambient…+95 °C Ambient…+95 °C Ambient…+95 °C Ambient…+95 °C Ambient…+95 °C Ambient…+95 °C Ambient…+95 °C Ambient…+95 °C Ambient…+95 °C Ambient…+95 °C Ambient…+95 °C Ambient…+95 °C Ambient…+95 °C Ambient…+95 °C Ambient…+95 °C Ambient…+95 °C Ambient…+95 °C Ambient…+95 °C Ambient…+95 °C Ambient…+95 °C Ambient…+95 °C Ambient…+95 °C Ambient…+95 °C Ambient…+95 °C Ambient…+95 °C Ambient…+95 °C Ambient…+95 °C Ambient…+95 °C Ambient…+95 °C
W×D 270×400 mm 270×400 mm 270×400 mm 270×400 mm 270×400 mm 270×400 mm 270×400 mm 270×400 mm 270×400 mm 270×400 mm 270×400 mm 270×400 mm 270×400 mm 270×400 mm 270×400 mm 270×400 mm 270×400 mm 270×400 mm 270×400 mm 270×400 mm 270×400 mm 270×400 mm 270×400 mm 270×400 mm 270×400 mm 270×400 mm 270×400 mm 270×400 mm 270×400 mm 270×400 mm 270×400 mm 270×400 mm 270×400 mm 270×400 mm 270×400 mm 270×400 mm 270×400 mm