Accessories for KL, KM, KS, SM and VKS shakers
Supplier: Edmund Bühler GmbH
0052059 0052058 0052056 0052057 0052201
BUHL0052059EA 174 SGD
BUHL0052059 BUHL0052058 BUHL0052056 BUHL0052057 BUHL0052201
Accessories for KL, KM, KS, SM and VKS shakers
Shakers Orbital Shakers
The test tube racks can be fastened either on the standard rack system between the clamping strips or can be screwed on the universal tray by means of a hinged foot. Hinged foot is used to vary the angle of inclination of the test tubes. The microplate is tilted at approximately 20°