Infrared heaters, IRB series
Supplier: Edmund Bühler GmbH
6063000 6060000 9066000
BUHL6063000EA 1950 SGD
BUHL6063000 BUHL6060000 BUHL9066000
Infrared heaters, IRB series
Heaters Hotplates
The IRB infrared heaters heated up samples to 700 or 900 °C within a short time.
  • Ideal for use where unshielded flames are not allowed for safety reasons