Orbital/reciprocating shakers, SM 30 A and SM 30 A control / SM 30 C and SM 30 C control
Supplier: Edmund Bühler GmbH
6101000 6104000 6105000 6100000
BUHL6101000EA 9520 SGD
BUHL6101000 BUHL6104000 BUHL6105000 BUHL6100000
Orbital/reciprocating shakers, SM 30 A and SM 30 A control / SM 30 C and SM 30 C control
Shakers Orbital Shakers
Benchtop orbital shakers which are suitable for continuous operation and accommodate loads of up to 30 kg. 'A' models are orbital motion only, the versatile 'C' models have functionality to change motion from orbital to reciprocating without using any tools. Universal tray accepts a maximum of 11×1000 or 15×500 ml flask clamps, Combifix rack system accommodates 6×1000 or 12×500 ml Erlenmeyer flasks. Variety of different vessels can be accommodated by choosing from the range of optional attachments.
  • Motion of 'C' models can be changed from orbital to
More Product Information