Microplate shakers, TiMix 2
Supplier: Edmund Bühler GmbH
6110000
BUHL6110000EA 5500 SGD
BUHL6110000
Microplate shakers, TiMix 2
Shakers Microplate Shakers
The TiMix 2 microplate shaker is specially for small samples in microplates, deepwell-plates or other plates in microplate format.
  • Holds four microplates
  • Timer (0 to 120 min) or continuous mode
  • Steplessly adjustable electronic speed control
Certifications: IP protection class according to DIN EN 60529: IP 21
Ordering information: Supplied without attachments or trays.