Multi-flask orbital shaker, VKS 75 A control
Supplier: Edmund Bühler GmbH
6111000
BUHL6111000EA 23120 SGD
BUHL6111000
Multi-flask orbital shaker, VKS 75 A control
Shakers Orbital Shakers
Large, heavy duty orbital shaker for use as a floor standing unit only. Shakers are designed for continuous operation and accommodate loads of up to 75 kg. A variety of different vessels can be accommodated in different configurations by choosing from the wide range of optional attachments.
  • Digital display with a new programmable control with extended functionality
  • Continuous operation or programmable
  • Infinitely variable speed control
  • Can be used at 5 to 50 °C (85% relative humidity), in an incubator or cold room
  • Wide range of accessories available, and up
More Product Information