Multi-flask reciprocating shaker, VKS 75 B control
Supplier: Edmund Bühler GmbH
6112000
BUHL6112000EA 23500 SGD
BUHL6112000
Multi-flask reciprocating shaker, VKS 75 B control
Shakers Reciprocating Shakers
Large, heavy duty reciprocating shaker for use as a floor standing unit only. Shaker is designed for continuous operation and accommodates loads of up to 75 kg. A variety of different vessels can be accommodated in different configurations by choosing from the wide range of optional attachments.
  • Digital display with a new programmable control with extended functionality
  • Continuous operation or programmable
  • Infinitely variable speed control
  • Can be used at 5 to 50 °C (85% relative humidity), in a cold room
  • Wide range of accessories available, and up to six
More Product Information