Orbital shaker with heating plate, SM 30 AT control
Supplier: Edmund Bühler GmbH
6200000
BUHL6200000EA 17410 SGD
BUHL6200000
Orbital shaker with heating plate, SM 30 AT control
Shakers Orbital Shakers
Bench-top orbital shaker with heating plate which is suitable for mixing of difficult media. This shaker is the ideal solution when the use of an incubator hood is not possible due to the use of solvents (as solvents could destroy electrical parts and other materials of the incubator hood). The unit features digital display of set and actual values and programmable control.
  • Continuous operation or programmable
  • Infinitely variable speed control, and units can be used at 5 to 50 °C (85% relative humidity)
  • Temperature range up to 120 °C with ±1 °C control
More Product Information