Respiratory hoods, Tychem®, Bullard®
Supplier: BULLARD
20SIC 20SICH 20SICN 20TICH 20TICN 20TICS 20TJ 20TJN RT1
BULL20SICEA 179.53 SGD
BULL20SIC BULL20SICH BULL20SICN BULL20TICH BULL20TICN BULL20TICS BULL20TJ BULL20TJN BULLRT1
Respiratory hoods, Tychem®, Bullard®
Respirators Respiratory Systems
Tychem® hoods provide a variety of simple, effective, and economical options for respiratory protection.
  • Simple, effective, and economical options for respiratory protection
  • Covers entire head, face, and neck to provide splash and overspray protection
  • Neck cuff design maintains positive pressure inside the hood by inflating the inner collar to provide a snug, comfortable fit and continuous airflow
  • Wide-angle lens offers large field of vision while accommodating beards, prescription eyeglasses, or safety glasses
  • Lighter and more comfortable than tight-
More Product Information