EVA PAPRs (powered air purifying respirators), complete systems
Supplier: BULLARD
EVA20TJ
BULLEVA20TJEA 2380.4 SGD
BULLEVA20TJ
EVA PAPRs (powered air purifying respirators), complete systems
Respirators Respiratory Systems
The EVA is simple, safe, and dependable.
  • Versatile respirator for particulate or chemical environments
  • Lightweight, compact, well-balanced design
  • Compatible with range of head tops
  • Run time: 4 to 10 hours
  • Belt type: standard comfort
In addition, the EVA offers Active Flow Technology™, which maintains an airflow of 8,5 CFM (high speed), 1000 Assigned Protection Factor (depending upon head top), and dual alarms for low flow and low battery. Finally, the EVA has a visual battery fuel gauge to check remaining charge at the push of a button, a brushlessMore Product Information