EVAHL PAPRs (powered air purifying respirators), blower assemblies
Supplier: BULLARD
EVAHL1 EVAHL2
BULLEVAHL1EA 1549.75 SGD
BULLEVAHL1 BULLEVAHL2
EVAHL PAPRs (powered air purifying respirators), blower assemblies
Respirators Respiratory Systems
The EVAHL is simple, safe, and dependable. The system features one-button operation, a choice of two air speeds, and a single filter design.
  • Certified for hazardous locations (Class I Division 2, Class II Division 2, Class III)
  • Versatile respirator for particulate or chemical environments
  • Lightweight, compact, well-balanced design
  • Run time: 4 to 10 hours
Additionally, the EVAHL offers Active Flow Technology™, which maintains an airflow of 8,5 CFM (high speed), 25/1000 Assigned Protection Factor (depending upon head top), and dual alarms for low flowMore Product Information