EVAHL PAPRs (powered air purifying respirators), complete systems
Supplier: BULLARD
EVAHL20LFL EVAHL20SICT EVAHL20TICT EVAHLRT3T EVAHLRT3TDB
BULLEVAHL20LFLEA 4690 SGD
BULLEVAHL20LFL BULLEVAHL20SICT BULLEVAHL20TICT BULLEVAHLRT3T BULLEVAHLRT3TDB
EVAHL PAPRs (powered air purifying respirators), complete systems
Respirators Respiratory Systems
The EVAHL is simple, safe, and dependable. The system features one-button operation, a choice of two air speeds, and a single filter design.
  • Certified for hazardous locations (Class I Division 2, Class II Division 2, Class III)
  • Versatile respirator for particulate or chemical environments
  • Lightweight, compact, well-balanced design
  • Run time: 4 to 10 hours
In addition, the EVAHL offers Active Flow Technology™, which maintains an airflow of 8,5 CFM (high speed), 1000 Assigned Protection Factor (depending upon headtop), and dual alarms for low flow andMore Product Information