Tamper-evident closure for compact jerrycan
Supplier: BURKLE
0300-0000
BURK0300-0000EA 4.5 SGD
BURK0300-0000
Tamper-evident closure for compact jerrycan
Closures Safety Caps and Safety Waste Caps
Screw-on closure with tamper-evident ring for compact jerrycan.
  • Screw-on cover made of PP
  • Seal made out of PE
  • Thread DIN 50
The closure offers protection against tampering and a guarantee that the container has not been opened before, since the snap ring is visibly broken the first time the container is opened.