Spray bottles, LaboPlast®
Supplier: BURKLE
0309-1100 0309-1025 0309-1050
BURK0309-1100EA 19.7 SGD
BURK0309-1100 BURK0309-1025 BURK0309-1050
Spray bottles, LaboPlast®
Bottles Spray Bottles
LDPE, translucent, PP spray attachment.
  • Good chemical resistance, good temperature resistance
  • Spray nozzle adjustment can be varied by rotation
  • Pressureless spray bottle
  • Media-contacting parts PP, PE and stainless steel
Spray, choice of fine mist or focused jet spray functions to disperse fluids.