Spigot, brass
Supplier: BURKLE
0535-0010
BURK0535-0010EA 122 SGD
BURK0535-0010
Spigot, brass
Stopcocks
Barrel tap for petrochemical liquids.
  • Seals NBR
  • NW 10 mm
  • Closed independently when not used (increased safety)